Gene Transcripts
- Gene Symbol
- ELP6
- Entrez Gene ID
- 54859
- Gene Name
- elongator acetyltransferase complex subunit 6
- Gene Aliases
- C3orf75, TMEM103
- Chromosome Location
- Chr. 3: 47495640 - 47513711 on Build GRCh38
- UniGene ID
- Hs.311100
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031703.2 |
NP_001026873.2
|
2, 3 |
262 |
|
XM_011533851.2 |
XP_011532153.1
|
2, 3 |
264 |
|
XM_011533852.2 |
XP_011532154.1
|
2, 3 |
600 |
|
XM_011533853.2 |
XP_011532155.1
|
2, 3 |
241 |
|
XM_017006669.1 |
XP_016862158.1
|
2, 3 |
264 |
|
XM_017006670.1 |
XP_016862159.1
|
2, 3 |
533 |
|
GenBank mRNA |
AK000218.1 |
- |
2, 3 |
247 |
|
AK225721.1 |
- |
2, 3 |
244 |
|
BC000623.2 |
- |
2, 3 |
166 |
3346325 |
BC014204.2 |
- |
2, 3 |
166 |
4620013 |
DQ787088.1 |
- |
2, 3 |
143 |
|
View Full Product Details