Gene Transcripts

Gene Symbol
DPP8
Entrez Gene ID
54878
Gene Name
dipeptidyl peptidase 8
Gene Aliases
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
Chromosome Location
Chr. 15: 65442463 - 65517713 on Build GRCh38
UniGene ID
Hs.458609
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320875.1 NP_001307804.1 5 881
NM_001320876.1 NP_001307805.1 4 850
NM_017743.5 NP_060213.2 4 745
NM_130434.4 NP_569118.1 4 745
NM_197960.3 NP_932064.1 5 786
NM_197961.3 NP_932065.1 5 881
NR_135485.1 4 745
NR_135486.1 5 786
XM_011521730.1 XP_011520032.1 4 2134
XM_011521732.2 XP_011520034.1 4 2134
XM_011521734.1 XP_011520036.1 4 2134
XM_017022373.1 XP_016877862.1 4 2134
XM_017022374.1 XP_016877863.1 4 2134
XM_017022375.1 XP_016877864.1 4 726
XM_017022376.1 XP_016877865.1 4 726
XM_017022378.1 XP_016877867.1 4 2134
GenBank mRNA AF221634.1 - 4 719
AK000290.1 - 4 668
AK291057.1 - 5 856
AK296915.1 - 4 821
AK302638.1 - 5 722
AY172659.1 - 3 513
AY354202.1 - 5 714
BC030688.2 - 5 872 4822550

View Full Product Details