Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042479.1 | NP_001035944.1 | 4 | 421 | |
NM_001042480.1 | NP_001035945.1 | 3 | 339 | ||
NM_017856.2 | NP_060326.1 | 4 | 509 | ||
XM_005274555.2 | XP_005274612.1 | 3 | 3374 | ||
XM_017029617.1 | XP_016885106.1 | 3 | 510 | ||
XM_017029618.1 | XP_016885107.1 | 3 | 510 | ||
GenBank mRNA | AK000521.1 | - | 4 | 486 | |
AK125062.1 | - | 3 | 2466 | ||
BC003607.2 | - | 3 | 263 | 2987964 | |
BC014324.1 | - | 3 | 320 | 3930643 | |
BC020785.1 | - | 3 | 316 | 4808527 | |
BX386466.2 | - | 4 | 387 | ||
CR457241.1 | - | 1 | 166 | ||
DQ224033.1 | - | 1 | 166 |