Gene Transcripts

Gene Symbol
EMC3
Entrez Gene ID
55831
Gene Name
ER membrane protein complex subunit 3
Gene Aliases
POB, TMEM111
Chromosome Location
Chr. 3: 9962680 - 10011248 on Build GRCh38
UniGene ID
Hs.475392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018447.3 NP_060917.1 2, 3 625
XM_005265321.3 XP_005265378.1 2, 3 493
GenBank mRNA AF157321.1 - 1, 2 337
AK172843.1 - 1, 2 333
AK222953.1 - 1, 2 335
AK312008.1 - 1, 2 332
BC022807.1 - 1, 2 320 4897577
DA359966.1 - 2, 3 493

View Full Product Details