Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
56942
Gene Name
C-X9-C motif containing 2
Gene Aliases
2310061C15Rik, C16orf61, DC13
Chromosome Location
Chr. 16: 80975800 - 81007046 on Build GRCh38
UniGene ID
Hs.55028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020188.3 NP_064573.1 3, 4 348
XM_017023461.1 XP_016878950.1 4, 5 536
XM_017023467.1 XP_016878956.1 5, 6 431
XM_017023468.1 XP_016878957.1 4, 5 341
XM_017023469.1 XP_016878958.1 4, 5 415
XM_017023470.1 XP_016878959.1 4, 5 339
GenBank mRNA AF201935.1 - 3, 4 324
AK123193.1 - 2, 3 1703
BC032631.1 - 3, 4 352 5399193

View Full Product Details