Gene Transcripts

Gene Symbol
IGSF9
Entrez Gene ID
57549
Gene Name
immunoglobulin superfamily member 9
Gene Aliases
FP18798, IGSF9A, Nrt1
Chromosome Location
Chr. 1: 159927039 - 159945661 on Build GRCh38
UniGene ID
Hs.591472
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135050.1 NP_001128522.1 19 3089
NM_020789.3 NP_065840.2 19 3041
GenBank mRNA AB037776.1 - 19 3089
AK056650.1 - 8 3250
AY203932.1 - 7 2607
AY358128.1 - 12 1721
BC030141.2 - 19 3041 4762143

View Full Product Details