Gene Transcripts

Gene Symbol
MICAL3
Entrez Gene ID
57553
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 3
Gene Aliases
MICAL-3
Chromosome Location
Chr. 22: 17787649 - 18024559 on Build GRCh38
UniGene ID
Hs.528024
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122731.2 NP_001116203.1 2 376
NM_001136004.3 NP_001129476.1 2 376
NM_015241.2 NP_056056.2 3 655
XM_005261262.3 XP_005261319.1 3 485
XM_011546137.2 XP_011544439.1 3 485
XM_011546139.2 XP_011544441.1 3 485
XM_011546142.2 XP_011544444.1 3 485
XM_011546146.2 XP_011544448.1 3 485
XM_017028865.1 XP_016884354.1 3 1003
XM_017028866.1 XP_016884355.1 3 809
XM_017028867.1 XP_016884356.1 3 1926
XM_017028868.1 XP_016884357.1 3 655
XM_017028869.1 XP_016884358.1 3 400
XM_017028870.1 XP_016884359.1 3 401
XM_017028871.1 XP_016884360.1 3 485
XM_017028872.1 XP_016884361.1 3 485
XM_017028873.1 XP_016884362.1 3 485
XM_017028874.1 XP_016884363.1 3 485
XM_017028875.1 XP_016884364.1 3 485
XM_017028876.1 XP_016884365.1 3 485
XM_017028877.1 XP_016884366.1 3 485
XM_017028878.1 XP_016884367.1 3 485
XM_017028879.1 XP_016884368.1 3 485
XM_017028880.1 XP_016884369.1 3 808
XM_017028881.1 XP_016884370.1 3 655
XM_017028882.1 XP_016884371.1 3 485
XM_017028883.1 XP_016884372.1 1 40
XM_017028884.1 XP_016884373.1 3 826
XM_017028887.1 XP_016884376.1 3 485
GenBank mRNA AK127487.1 - 3 485
AK295978.1 - 3 627
BC157876.1 - 2 364 8862700
BC171887.1 - 2 364 9054541
BX647382.1 - 2 325
CR456364.1 - 2 317

View Full Product Details