Gene Transcripts

Gene Symbol
CHP2
Entrez Gene ID
63928
Gene Name
calcineurin like EF-hand protein 2
Gene Aliases
Chromosome Location
Chr. 16: 23754627 - 23758951 on Build GRCh38
UniGene ID
Hs.178589
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022097.3 NP_071380.1 5 819
GenBank mRNA AF146019.1 - 5 824
AK290253.1 - 5 405
AK313062.1 - 5 675

View Full Product Details