Gene Transcripts

Gene Symbol
TMEM267
Entrez Gene ID
64417
Gene Name
transmembrane protein 267
Gene Aliases
C5orf28
Chromosome Location
Chr. 5: 43444252 - 43484660 on Build GRCh38
UniGene ID
Hs.732093
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022483.4 NP_071928.2 2 242
XM_005248333.4 XP_005248390.1 3 1110
XM_005248335.3 XP_005248392.1 3 394
XM_006714486.3 XP_006714549.1 2 714
XM_011514074.2 XP_011512376.1 4 1486
XM_011514075.2 XP_011512377.1 4 1482
GenBank mRNA AK025310.1 - 2 211
AK225220.1 - 2 211
AK315466.1 - 3 394
BC013351.2 - 3 431 3621124
BC063832.1 - 1 188 6178186
BC119658.1 - 1 148 40115985
BC119659.1 - 1 148 40115986
DA028175.1 - 2 242
DA035926.1 - 2 211

View Full Product Details