Gene Transcripts
- Gene Symbol
- TTC31
- Entrez Gene ID
- 64427
- Gene Name
- tetratricopeptide repeat domain 31
- Gene Aliases
- Chromosome Location
- Chr. 2: 74483032 - 74494564 on Build GRCh38
- UniGene ID
- Hs.557709
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022492.4 |
NP_071937.4
|
1, 2 |
60 |
|
NR_027749.1 |
|
1, 2 |
60 |
|
XM_005264492.1 |
XP_005264549.1
|
1, 2 |
49 |
|
XM_011533035.1 |
XP_011531337.1
|
1, 2 |
49 |
|
XM_011533036.1 |
XP_011531338.1
|
1, 2 |
49 |
|
XM_011533037.1 |
XP_011531339.1
|
1, 2 |
49 |
|
XM_011533040.1 |
XP_011531342.1
|
1, 2 |
101 |
|
XM_017004706.1 |
XP_016860195.1
|
1, 2 |
49 |
|
XM_017004707.1 |
XP_016860196.1
|
1, 2 |
49 |
|
XM_017004708.1 |
XP_016860197.1
|
1, 2 |
57 |
|
GenBank mRNA |
AK022850.1 |
- |
1, 2 |
46 |
|
AK090520.1 |
- |
1, 2 |
49 |
|
AK223355.1 |
- |
1, 2 |
44 |
|
AK225626.1 |
- |
1, 2 |
46 |
|
AK301585.1 |
- |
1, 2 |
44 |
|
AK303105.1 |
- |
1, 2 |
46 |
|
AK303245.1 |
- |
1, 2 |
46 |
|
BC036017.1 |
- |
1, 2 |
44 |
5270575 |
BC045536.1 |
- |
1, 2 |
51 |
|
BC096709.1 |
- |
1, 2 |
46 |
40023059 |
BC098365.1 |
- |
1, 2 |
46 |
40023060 |
BC099729.1 |
- |
1, 2 |
46 |
|
BX648131.1 |
- |
1, 2 |
45 |
|
View Full Product Details