Gene Transcripts

Gene Symbol
FNDC4
Entrez Gene ID
64838
Gene Name
fibronectin type III domain containing 4
Gene Aliases
FRCP1
Chromosome Location
Chr. 2: 27491883 - 27495303 on Build GRCh38
UniGene ID
Hs.27836
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022823.2 NP_073734.1 6 970
GenBank mRNA AK026015.1 - 6 930
AY358136.1 - 6 688
BC032725.1 - 5 601 5533159

View Full Product Details