Gene Transcripts

Gene Symbol
TMEM43
Entrez Gene ID
79188
Gene Name
transmembrane protein 43
Gene Aliases
ARVC5, ARVD5, EDMD7, LUMA
Chromosome Location
Chr. 3: 14124940 - 14143680 on Build GRCh38
UniGene ID
Hs.517817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024334.2 NP_077310.1 2 293
XM_017007176.1 XP_016862665.1 3 415
GenBank mRNA AK027757.1 - 2 219
AK027877.1 - 2 157
AK075010.1 - 2 144
AK225589.1 - 2 216
AL136916.1 - 2 187
AY358625.1 - 2 56
BC003125.1 - 2 188 3503636
BC011719.2 - 2 188 4079766
DA259919.1 - 2 298
KU178769.1 - 1 39
KU178770.1 - 1 39

View Full Product Details