Gene Transcripts

Gene Symbol
OR5H6
Entrez Gene ID
79295
Gene Name
olfactory receptor family 5 subfamily H member 6 (gene/pseudogene)
Gene Aliases
OR3-11
Chromosome Location
Chr. 3: 98263260 - 98265262 on Build GRCh38
UniGene ID
Hs.554712
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005479.1 NP_001005479.1 1 335
XM_011513118.2 XP_011511420.1 2 381
GenBank mRNA BC137523.1 - 1 376
BC137524.1 - 1 376 9021524

View Full Product Details