Gene Transcripts

Gene Symbol
ZMAT4
Entrez Gene ID
79698
Gene Name
zinc finger matrin-type 4
Gene Aliases
Chromosome Location
Chr. 8: 40530590 - 40897828 on Build GRCh38
UniGene ID
Hs.591850
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135731.1 NP_001129203.1 5 554
NM_024645.2 NP_078921.1 6 782
GenBank mRNA AK023904.1 - 6 782
BC019598.1 - 5 540 4934088

View Full Product Details