Gene Transcripts

Gene Symbol
NAA40
Entrez Gene ID
79829
Gene Name
N(alpha)-acetyltransferase 40, NatD catalytic subunit
Gene Aliases
NAT11, PATT1
Chromosome Location
Chr. 11: 63938970 - 63957322 on Build GRCh38
UniGene ID
Hs.523753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300800.1 NP_001287729.1 5 486
NM_024771.3 NP_079047.2 5 415
XM_006718689.3 XP_006718752.1 5 402
XM_011545254.2 XP_011543556.1 6 489
XM_011545255.2 XP_011543557.1 6 489
GenBank mRNA AK023910.1 - 5 384
AK299041.1 - 5 480
AK300451.1 - 4 336
BC041617.1 - 5 344 5921480
BC052298.1 - 5 344 6292760
BX647309.1 - 4 1441
DA649381.1 - 5 420
HY009487.1 - 5 486
JF263566.1 - 5 384

View Full Product Details