Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001304957.1 | NP_001291886.1 | 5 | 1098 | |
NM_001304958.1 | NP_001291887.1 | 6 | 1205 | ||
NM_001304959.1 | NP_001291888.1 | 5 | 847 | ||
NM_001304960.1 | NP_001291889.1 | 7 | 1016 | ||
NM_001304961.1 | NP_001291890.1 | 4 | 1015 | ||
NM_025140.2 | NP_079416.1 | 5 | 1108 | ||
XM_005253624.2 | XP_005253681.1 | 5 | 852 | ||
XM_017019984.1 | XP_016875473.1 | 5 | 1666 | ||
GenBank mRNA | AB015292.1 | - | 4 | 628 | |
AK026124.1 | - | 5 | 739 | ||
AK054680.1 | - | 3 | 2375 | ||
AK124358.1 | - | 1 | 2197 | ||
AK125866.1 | - | 3 | 2626 | ||
AK222661.1 | - | 5 | 696 | ||
BC017186.2 | - | 5 | 731 | 3051346 | |
BX394686.2 | - | 4 | 763 |