Gene Transcripts

Gene Symbol
ACAD10
Entrez Gene ID
80724
Gene Name
acyl-CoA dehydrogenase family member 10
Gene Aliases
Chromosome Location
Chr. 12: 111686053 - 111757107 on Build GRCh38
UniGene ID
Hs.331141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136538.1 NP_001130010.1 21 3317
NM_025247.5 NP_079523.3 20 3224
GenBank mRNA AK097425.1 - 13 1950
AL832043.1 - 21 3295
AY323912.1 - 20 3180
BC003698.1 - 16 2381
BC126358.1 - 20 3138 8992074

View Full Product Details