Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030641.3 | NP_085144.1 | 2 | 329 | |
XM_011530392.2 | XP_011528694.1 | 3 | 506 | ||
GenBank mRNA | AK095881.1 | - | 2 | 291 | |
AL834420.1 | - | 2 | 293 | ||
AX775805.1 | - | 2 | 301 | ||
AY014879.1 | - | 2 | 329 | ||
AY358210.1 | - | 2 | 226 | ||
BC038950.2 | - | 2 | 259 | 5552006 | |
BC047864.1 | - | 2 | 311 | 5498605 | |
BU585026.1 | - | 2 | 339 | ||
CR456381.1 | - | 1 | 61 |