Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001257137.2 | NP_001244066.1 | 25 | 2771 | |
NM_001257138.2 | NP_001244067.1 | 23 | 2506 | ||
NM_001324197.1 | NP_001311126.1 | 25 | 2792 | ||
NM_001324198.1 | NP_001311127.1 | 24 | 2669 | ||
NM_031483.6 | NP_113671.3 | 24 | 2648 | ||
XM_005260570.1 | XP_005260627.1 | 23 | 2564 | ||
XM_011529079.1 | XP_011527381.1 | 23 | 2584 | ||
XM_017028089.1 | XP_016883578.1 | 24 | 2604 | ||
XM_017028090.1 | XP_016883579.1 | 25 | 2724 | ||
XM_017028091.1 | XP_016883580.1 | 23 | 2519 | ||
GenBank mRNA | AB056663.2 | - | 24 | 2558 | |
AB209747.1 | - | 16 | 1665 | ||
AF038564.1 | - | 19 | 2066 | ||
AF095745.1 | - | 22 | 2441 | ||
AK297809.1 | - | 24 | 2466 | ||
AK304090.1 | - | 23 | 2465 | ||
AK315212.1 | - | 24 | 2607 | ||
BC006848.1 | - | 3 | 204 | ||
BC011571.1 | - | 25 | 2694 | 4560366 |