Gene Transcripts

Gene Symbol
BEX2
Entrez Gene ID
84707
Gene Name
brain expressed X-linked 2
Gene Aliases
BEX1, DJ79P11.1
Chromosome Location
Chr. X: 103309346 - 103311046 on Build GRCh38
UniGene ID
Hs.398989
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168399.1 NP_001161871.1 3 649
NM_001168400.1 NP_001161872.1 3 646
NM_001168401.1 NP_001161873.1 3 391
NM_032621.3 NP_116010.1 3 463
GenBank mRNA AF251053.1 - 3 295
AK312085.1 - 3 387
AY833560.1 - 3 395
BC015522.1 - 3 341 3914401
BU569325.1 - 3 466
BU596902.1 - 3 374
CB961647.1 - 3 655
CD365521.1 - 1 448
CK001963.1 - 3 394

View Full Product Details