Gene Transcripts
- Gene Symbol
- NLRP12
- Entrez Gene ID
- 91662
- Gene Name
- NLR family pyrin domain containing 12
- Gene Aliases
- CLR19.3, FCAS2, NALP12, PAN6, PYPAF7, RNO, RNO2
- Chromosome Location
- Chr. 19: 53793584 - 53824403 on Build GRCh38
- UniGene ID
- Hs.631573
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001277126.1 |
NP_001264055.1
|
5, 6 |
2641 |
|
NM_001277129.1 |
NP_001264058.1
|
5, 6 |
2638 |
|
NM_144687.3 |
NP_653288.1
|
5, 6 |
2638 |
|
XM_011527480.1 |
XP_011525782.1
|
5, 6 |
2641 |
|
XM_011527482.1 |
XP_011525784.1
|
5, 6 |
2641 |
|
XM_017027460.1 |
XP_016882949.1
|
5, 6 |
2641 |
|
XM_017027461.1 |
XP_016882950.1
|
5, 6 |
2641 |
|
XM_017027462.1 |
XP_016882951.1
|
4, 5 |
2467 |
|
XM_017027463.1 |
XP_016882952.1
|
6, 7 |
2414 |
|
XM_017027464.1 |
XP_016882953.1
|
5, 6 |
2212 |
|
XM_017027465.1 |
XP_016882954.1
|
6, 7 |
2422 |
|
XM_017027466.1 |
XP_016882955.1
|
6, 7 |
2419 |
|
XM_017027467.1 |
XP_016882956.1
|
5, 6 |
2214 |
|
GenBank mRNA |
AF231021.1 |
- |
3, 4 |
1566 |
|
AK095460.1 |
- |
5, 6 |
2553 |
|
AK290772.1 |
- |
5, 6 |
2420 |
|
AY095146.1 |
- |
5, 6 |
2585 |
|
AY116204.1 |
- |
5, 6 |
2632 |
|
AY116205.1 |
- |
5, 6 |
2632 |
|
AY116206.1 |
- |
5, 6 |
2632 |
|
AY116207.1 |
- |
5, 6 |
2632 |
|
AY154467.1 |
- |
5, 6 |
2585 |
|
BC028069.1 |
- |
5, 6 |
2528 |
5212737 |
View Full Product Details