Gene Transcripts
- Gene Symbol
- FRA10AC1
- Entrez Gene ID
- 118924
- Gene Name
- fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
- Gene Aliases
- C10orf4, F26C11.1-like, FRA10A
- Chromosome Location
- Chr. 10: 93667883 - 93702572 on Build GRCh38
- UniGene ID
- Hs.586650
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145246.4 |
NP_660289.2
|
6, 7 |
582 |
|
XM_005269515.4 |
XP_005269572.1
|
7, 8 |
679 |
|
XM_005269516.4 |
XP_005269573.1
|
6, 7 |
774 |
|
XM_011539259.2 |
XP_011537561.1
|
6, 7 |
547 |
|
XM_011539260.2 |
XP_011537562.1
|
7, 8 |
637 |
|
XM_017015656.1 |
XP_016871145.1
|
6, 7 |
547 |
|
XM_017015657.1 |
XP_016871146.1
|
6, 7 |
582 |
|
XM_017015658.1 |
XP_016871147.1
|
7, 8 |
682 |
|
XM_017015659.1 |
XP_016871148.1
|
6, 7 |
774 |
|
XM_017015660.1 |
XP_016871149.1
|
7, 8 |
639 |
|
XM_017015661.1 |
XP_016871150.1
|
7, 8 |
696 |
|
XM_017015662.1 |
XP_016871151.1
|
6, 7 |
547 |
|
GenBank mRNA |
AJ431721.1 |
- |
6, 7 |
578 |
|
AJ431722.1 |
- |
6, 7 |
578 |
|
AJ431723.1 |
- |
6, 7 |
578 |
|
AJ431724.1 |
- |
6, 7 |
578 |
|
AJ431725.1 |
- |
6, 7 |
578 |
|
AK090955.1 |
- |
6, 7 |
582 |
|
BC018007.1 |
- |
6, 7 |
451 |
4278022 |
BM272089.1 |
- |
6, 7 |
566 |
|
BX648699.1 |
- |
5, 6 |
616 |
|
View Full Product Details