Gene Transcripts

Gene Symbol
CHMP4B
Entrez Gene ID
128866
Gene Name
charged multivesicular body protein 4B
Gene Aliases
C20orf178, CHMP4A, CTPP3, CTRCT31, SNF7, SNF7-2, Shax1, VPS32B, Vps32-2, dJ553F4.4
Chromosome Location
Chr. 20: 33811304 - 33854367 on Build GRCh38
UniGene ID
Hs.472471
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176812.4 NP_789782.1 3, 4 658
GenBank mRNA AB100261.1 - 3, 4 603
AY329085.1 - 3, 4 493
BC033859.1 - 3, 4 602 5173394

View Full Product Details