Gene Transcripts
- Gene Symbol
- IGFL2
- Entrez Gene ID
- 147920
- Gene Name
- IGF like family member 2
- Gene Aliases
- UNQ645, VPRI645
- Chromosome Location
- Chr. 19: 46078513 - 46195097 on Build GRCh38
- UniGene ID
- Hs.99376
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002915.2 |
NP_001002915.2
|
3, 4 |
391 |
|
NM_001135113.1 |
NP_001128585.1
|
2, 3 |
103 |
|
XM_011526501.1 |
XP_011524803.1
|
3, 4 |
2930 |
|
XM_017026335.1 |
XP_016881824.1
|
2, 3 |
779 |
|
GenBank mRNA |
AK093330.1 |
- |
1, 2 |
1417 |
|
AK125754.1 |
- |
3, 4 |
1249 |
|
AY358692.1 |
- |
2, 3 |
103 |
|
AY672112.1 |
- |
3, 4 |
391 |
|
BC130490.1 |
- |
2, 3 |
192 |
40146521 |
BC130492.1 |
- |
2, 3 |
192 |
40146523 |
View Full Product Details