Gene Transcripts

Gene Symbol
GDF7
Entrez Gene ID
151449
Gene Name
growth differentiation factor 7
Gene Aliases
BMP12
Chromosome Location
Chr. 2: 20666664 - 20671490 on Build GRCh38
UniGene ID
Hs.447688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182828.3 NP_878248.2 2 1877
GenBank mRNA AB158468.1 - 2 1359
AF522369.1 - 2 1877

View Full Product Details