Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207352.3 | NP_997235.3 | 2 | 539 | |
XM_005262935.3 | XP_005262992.1 | 2 | 324 | ||
XM_017008037.1 | XP_016863526.1 | 2 | 383 | ||
GenBank mRNA | AK122600.1 | - | 2 | 274 | |
AK126473.1 | - | 3 | 208 | ||
AY422002.1 | - | 2 | 539 | ||
BC060857.1 | - | 2 | 405 | 30333559 |