Gene Transcripts

Gene Symbol
TGM6
Entrez Gene ID
343641
Gene Name
transglutaminase 6
Gene Aliases
SCA35, TG6, TGM3L, TGY, dJ734P14.3
Chromosome Location
Chr. 20: 2380908 - 2432753 on Build GRCh38
UniGene ID
Hs.452039
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001254734.1 NP_001241663.1 10 1695
NM_198994.2 NP_945345.2 10 1695
GenBank mRNA AF540969.1 - 10 1695
AF540970.1 - 10 1695

View Full Product Details