Gene Transcripts

Gene Symbol
C15orf53
Entrez Gene ID
400359
Gene Name
chromosome 15 open reading frame 53
Gene Aliases
Chromosome Location
Chr. 15: 38696598 - 38700038 on Build GRCh38
UniGene ID
Hs.448785
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207444.2 NP_997327.1 1 75
GenBank mRNA AK093014.1 - 1 75
BC119016.1 - 1 75 40108911
BC119810.1 - 1 75 40108916

View Full Product Details