Gene Transcripts

Gene Symbol
CST5
Entrez Gene ID
1473
Gene Name
cystatin D
Gene Aliases
Chromosome Location
Chr. 20: 23875935 - 23879743 on Build GRCh38
UniGene ID
Hs.121489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001900.4 NP_001891.2 3 475
GenBank mRNA BC062678.1 - 3 472
BC069514.1 - 3 412 7262733
X70377.1 - 3 412

View Full Product Details