Gene Transcripts

Gene Symbol
MARCKS
Entrez Gene ID
4082
Gene Name
myristoylated alanine rich protein kinase C substrate
Gene Aliases
80K-L, MACS, PKCSL, PRKCSL
Chromosome Location
Chr. 6: 113857335 - 113863475 on Build GRCh38
UniGene ID
Hs.519909
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002356.6 NP_002347.5 2 532
GenBank mRNA BC013004.2 - 2 505
BC028284.1 - 2 505
BC068059.1 - 2 498
BC089040.1 - 2 379 6378349
CN345330.1 - 2 507
D10522.1 - 2 490
M68956.1 - 2 429

View Full Product Details