Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002590.3 | NP_002581.2 | 1 | 1603 | |
NM_032949.2 | NP_116567.1 | 1 | 1603 | ||
GenBank mRNA | AF061573.2 | - | 1 | 1602 | |
AY013873.1 | - | 1 | 1399 | ||
BC036025.1 | - | 1 | 1603 | 5278690 |