Gene Transcripts

Gene Symbol
SNAI1
Entrez Gene ID
6615
Gene Name
snail family transcriptional repressor 1
Gene Aliases
SLUGH2, SNA, SNAH, SNAIL, SNAIL1, dJ710H13.1
Chromosome Location
Chr. 20: 49982976 - 49988886 on Build GRCh38
UniGene ID
Hs.48029
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005985.3 NP_005976.2 2 657
GenBank mRNA AF125377.1 - 2 634
AK313228.1 - 2 653
BC012910.1 - 2 638 4537122
BI822180.1 - 2 643

View Full Product Details