Gene Transcripts
- Gene Symbol
- SNORD22
- Entrez Gene ID
- 9304
- Gene Name
- small nucleolar RNA, C/D box 22
- Gene Aliases
- RNU22, U22
- Chromosome Location
- Chr. 11: 62852910 - 62853035 on Build GRCh38
- UniGene ID
- Hs.744737
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_000008.2 |
|
|
11 |
|
- Gene Symbol
- SNHG1
- Entrez Gene ID
- 23642
- Gene Name
- small nucleolar RNA host gene 1
- Gene Aliases
- LINC00057, NCRNA00057, U22HG, UHG
- Chromosome Location
- Chr. 11: 62851988 - 62855888 on Build GRCh38
- UniGene ID
- Hs.655643
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095849.1 |
- |
4 |
1861 |
|
View Full Product Details