Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006713.3 | NP_006704.3 | 5 | 474 | |
XM_011513944.2 | XP_011512246.1 | 6 | 727 | ||
XM_017008986.1 | XP_016864475.1 | 6 | 570 | ||
XM_017008987.1 | XP_016864476.1 | 6 | 447 | ||
GenBank mRNA | AB209285.1 | - | 5 | 369 | |
AK311754.1 | - | 5 | 416 | ||
BC009610.1 | - | 5 | 402 | 4297490 | |
BC010537.1 | - | 5 | 377 | 3457167 | |
BC018189.1 | - | 5 | 396 | 3881373 | |
BC022339.1 | - | 5 | 407 | 4134117 | |
BC063402.1 | - | 6 | 692 | ||
BC071655.1 | - | 5 | 391 | ||
BC105972.1 | - | 5 | 372 | 3907729 | |
BX472106.1 | - | 5 | 475 | ||
BX537584.1 | - | 2 | 4197 | ||
CR456902.1 | - | 4 | 346 |