Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012452.2 | NP_036584.1 | 3 | 264 | |
GenBank mRNA | AF023614.1 | - | 3 | 264 | |
AK223453.1 | - | 3 | 264 | ||
AK301032.1 | - | 3 | 254 | ||
AK313302.1 | - | 3 | 284 | ||
AX766384.1 | - | 3 | 264 | ||
AX772740.1 | - | 2 | 118 | ||
AY302137.1 | - | 2 | 113 | ||
BC109392.1 | - | 2 | 125 | 40030781 |