Gene Transcripts

Gene Symbol
KLF13
Entrez Gene ID
51621
Gene Name
Kruppel like factor 13
Gene Aliases
BTEB3, FKLF2, NSLP1, RFLAT-1, RFLAT1
Chromosome Location
Chr. 15: 31326855 - 31435665 on Build GRCh38
UniGene ID
Hs.376443
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302461.1 NP_001289390.1 1 529
NM_015995.3 NP_057079.2 1 529
GenBank mRNA AF132599.1 - 1 543
AF150628.1 - 1 219
BC010438.1 - 1 484 2823301
BC012741.2 - 1 387 3940815
BC013946.2 - 1 484 2823301
BC035503.2 - 1 504
KJ534876.1 - 1 171

View Full Product Details