Gene Transcripts

Gene Symbol
FUNDC2
Entrez Gene ID
65991
Gene Name
FUN14 domain containing 2
Gene Aliases
DC44, HCBP6, HCC3, PD03104
Chromosome Location
Chr. X: 155026789 - 155056916 on Build GRCh38
UniGene ID
Hs.356050
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023934.3 NP_076423.2 5 2523
GenBank mRNA BM713884.1 - 1 526
BU738737.1 - 1 455

View Full Product Details