Gene Transcripts

Gene Symbol
KRTAP4-3
Entrez Gene ID
85290
Gene Name
keratin associated protein 4-3
Gene Aliases
KAP4.3
Chromosome Location
Chr. 17: 41167231 - 41168172 on Build GRCh38
UniGene ID
Hs.307023
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033187.1 NP_149443.1 1 462
GenBank mRNA AJ406935.1 - 1 171

View Full Product Details