Gene Transcripts

Gene Symbol
PAGE5
Entrez Gene ID
90737
Gene Name
PAGE family member 5
Gene Aliases
CT16, CT16.1, CT16.2, GAGEE1, PAGE-5
Chromosome Location
Chr. X: 55220347 - 55224108 on Build GRCh38
UniGene ID
Hs.668491
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013435.2 NP_001013453.1 2, 3 184
NM_130467.4 NP_569734.2 2, 3 383
GenBank mRNA AJ344352.1 - 2, 3 353
BC009230.2 - 2, 3 372 3955765
BC110803.1 - 2, 3 118 3922538
BP429046.1 - 2, 3 188
BQ437151.1 - 2, 3 172

View Full Product Details