Gene Transcripts
- Gene Symbol
- CIDECP
- Entrez Gene ID
- 152302
- Gene Name
- cell death-inducing DFFA-like effector c pseudogene
- Gene Aliases
- CICE
- Chromosome Location
- Chr. 3: 10017553 - 10026136 on Build GRCh38
- UniGene ID
- Hs.635072
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002786.1 |
|
3 |
566 |
|
View Full Product Details