Gene Transcripts
- Gene Symbol
- RBMY2FP
- Entrez Gene ID
- 159162
- Gene Name
- RNA binding motif protein, Y-linked, family 2, member F pseudogene
- Gene Aliases
- Chromosome Location
- Chr. Y: 22308859 - 22316205 on Build GRCh38
- UniGene ID
- Hs.711527
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002193.2 |
|
4 |
952 |
|
GenBank mRNA |
BC026077.2 |
- |
4 |
954 |
|
View Full Product Details