Gene Transcripts
- Gene Symbol
- COMMD6
- Entrez Gene ID
- 170622
- Gene Name
- COMM domain containing 6
- Gene Aliases
- Acrg
- Chromosome Location
- Chr. 13: 75525214 - 75549439 on Build GRCh38
- UniGene ID
- Hs.508266
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287392.1 |
NP_001274321.1
|
1, 2 |
99 |
|
NM_001287393.1 |
NP_001274322.1
|
1, 2 |
99 |
|
NM_203495.3 |
NP_987091.1
|
1, 2 |
99 |
|
NM_203497.3 |
NP_987093.1
|
1, 2 |
99 |
|
XM_011534948.2 |
XP_011533250.1
|
2, 3 |
192 |
|
GenBank mRNA |
AF169971.1 |
- |
1, 2 |
66 |
|
AK000009.1 |
- |
1, 2 |
68 |
|
AW516002.1 |
- |
4, 5 |
414 |
|
AY542161.1 |
- |
1, 2 |
32 |
|
BC110837.1 |
- |
1, 2 |
36 |
|
BC117391.1 |
- |
1, 2 |
76 |
40125942 |
BC143912.1 |
- |
1, 2 |
76 |
9052430 |
BC171758.1 |
- |
1, 2 |
76 |
9054411 |
BM476717.1 |
- |
|
32 |
|
BX538116.1 |
- |
1, 2 |
42 |
|
CD175553.1 |
- |
1, 2 |
99 |
|
DB487154.2 |
- |
1, 2 |
82 |
|
HY028175.1 |
- |
1, 2 |
72 |
|
HY045286.1 |
- |
1, 2 |
44 |
|
HY105279.1 |
- |
1, 2 |
100 |
|
View Full Product Details