Gene Transcripts

Gene Symbol
GRAMD1B
Entrez Gene ID
57476
Gene Name
GRAM domain containing 1B
Gene Aliases
Chromosome Location
Chr. 11: 123358299 - 123627771 on Build GRCh38
UniGene ID
Hs.144725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017018036.1 XP_016873525.1 1 110
XM_017018037.1 XP_016873526.1 2 409

View Full Product Details