Gene Transcripts

Gene Symbol
SUMF1
Entrez Gene ID
285362
Gene Name
sulfatase modifying factor 1
Gene Aliases
AAPA3037, FGE, UNQ3037
Chromosome Location
Chr. 3: 4034714 - 4467282 on Build GRCh38
UniGene ID
Hs.350475
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164674.1 NP_001158146.1 8 1926
NM_001164675.1 NP_001158147.1 8 1941
NM_182760.3 NP_877437.2 9 2001
GenBank mRNA AB448737.1 - 9 1968
AK057983.1 - 10 1896
AK075459.1 - 9 1979
AK094111.1 - 1 1688
BC017005.2 - 4 1240
BC110862.1 - 9 1972 5760490
CA413741.1 - 1 175

View Full Product Details