Gene Transcripts

Gene Symbol
IQCA1L
Entrez Gene ID
392843
Gene Name
IQ motif containing with AAA domain 1 like
Gene Aliases
IQCA1P1
Chromosome Location
Chr. 7: 151190873 - 151205784 on Build GRCh38
UniGene ID
Hs.649589
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304419.1 NP_001291348.1 17, 18 2148
GenBank mRNA AW444952.1 - 2, 3 469
DV080513.1 - 3, 4 474

View Full Product Details