Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_003561.2 | 18, 19 | 3165 | ||
NR_027768.1 | 16, 17 | 3380 | |||
GenBank mRNA | AK093485.1 | - | 10, 11 | 792 | |
AK131353.1 | - | 18, 19 | 3158 | ||
AL834175.1 | - | 16, 17 | 3133 | ||
BC045595.1 | - | 1, 2 | 83 | ||
BC150309.1 | - | 15, 16 | 2772 |