Gene Transcripts

Gene Symbol
ZNRF3
Entrez Gene ID
84133
Gene Name
zinc and ring finger 3
Gene Aliases
BK747E2.3, RNF203
Chromosome Location
Chr. 22: 28800657 - 29057488 on Build GRCh38
UniGene ID
Hs.604200
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011530438.2 XP_011528740.1 1 186
XM_017028990.1 XP_016884479.1 1 186

View Full Product Details