Gene Transcripts
- Gene Symbol
- MROH2A
- Entrez Gene ID
- 339766
- Gene Name
- maestro heat like repeat family member 2A
- Gene Aliases
- HEATR7B1
- Chromosome Location
- Chr. 2: 233774022 - 233833423 on Build GRCh38
- UniGene ID
- Hs.361778
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001287395.1 |
NP_001274324.1
|
12, 13 |
1432 |
|
XM_011511075.2 |
XP_011509377.1
|
12, 13 |
1609 |
|
XM_011511076.2 |
XP_011509378.1
|
12, 13 |
1609 |
|
XM_011511077.2 |
XP_011509379.1
|
12, 13 |
1609 |
|
XM_011511078.2 |
XP_011509380.1
|
12, 13 |
1609 |
|
XM_011511079.2 |
XP_011509381.1
|
12, 13 |
1609 |
|
XM_011511080.2 |
XP_011509382.1
|
12, 13 |
3080 |
|
XM_011511081.2 |
XP_011509383.1
|
12, 13 |
1609 |
|
XM_011511082.2 |
XP_011509384.1
|
12, 13 |
1609 |
|
XM_011511083.2 |
XP_011509385.1
|
12, 13 |
1609 |
|
XM_011511084.2 |
XP_011509386.1
|
12, 13 |
1609 |
|
XM_011511085.2 |
XP_011509387.1
|
6, 7 |
679 |
|
XM_011511086.2 |
XP_011509388.1
|
12, 13 |
1609 |
|
View Full Product Details