Gene Transcripts

Gene Symbol
PRCD
Entrez Gene ID
768206
Gene Name
progressive rod-cone degeneration
Gene Aliases
RP36
Chromosome Location
Chr. 17: 76527586 - 76555339 on Build GRCh38
UniGene ID
Hs.634380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077620.2 NP_001071088.1 3 308
NR_033357.1 3 379
XM_017025013.1 XP_016880502.1 3 2326
XM_017025014.1 XP_016880503.1 3 2326
XM_017025015.1 XP_016880504.1 3 2326
GenBank mRNA AK125617.1 - 3 379
BC043413.1 - 3 176
BC146898.1 - 3 249 9056826
BC146904.1 - 3 249 9056832
DQ390338.1 - 3 315

View Full Product Details