Gene Transcripts

Gene Symbol
KLF13
Entrez Gene ID
51621
Gene Name
Kruppel like factor 13
Gene Aliases
BTEB3, FKLF2, NSLP1, RFLAT-1, RFLAT1
Chromosome Location
Chr. 15: 31326855 - 31435665 on Build GRCh38
UniGene ID
Hs.376443
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302461.1 NP_001289390.1 2 1147
GenBank mRNA AI074707.1 - 1 84
BX096457.1 - 1 91

View Full Product Details